Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL22RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676847.3 | XP_005676904.2 | 2 - 3 | 361 | 69 | |
XM_018055813.1 | XP_017911302.1 | 2 - 3 | 364 | 69 | ||
XM_018055820.1 | XP_017911309.1 | 2 - 3 | 366 | 69 | ||
GenBank mRNA | KA344339.1 | - | 2 - 3 | 327 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |