Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL22RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676847.3 | XP_005676904.2 | 6 - 7 | 977 | 78 | |
XM_018055813.1 | XP_017911302.1 | 7 - 8 | 1058 | 78 | ||
XM_018055834.1 | XP_017911323.1 | 6 - 7 | 925 | 78 | ||
GenBank mRNA | KA344339.1 | - | 6 - 7 | 943 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |