Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679730.3 | XP_005679787.2 | 4 - 5 | 755 | 84 | |
XM_018047567.1 | XP_017903056.1 | 4 - 5 | 803 | 84 | ||
XM_018047568.1 | XP_017903057.1 | 4 - 5 | 653 | 84 | ||
GenBank mRNA | JO332210.2 | - | 3 - 4 | 304 | 84 | |
JO599841.2 | - | 2 - 3 | 835 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |