Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054380.1 | XP_017909869.1 | 5 - 6 | 1480 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |