Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A4RG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057500.1 | XP_017912989.1 | 5 - 6 | 754 | 96 | |
GenBank mRNA | JO512098.2 | - | 2 - 3 | 619 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |