Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690815.3 | XP_005690872.1 | 5 - 6 | 530 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |