Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLYCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061772.1 | XP_017917261.1 | 3 - 4 | 855 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |