Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695184.2 | XP_005695241.2 | 3 - 4 | 444 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |