Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065926.1 | XP_017921415.1 | 27 - 28 | 4877 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |