Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065926.1 | XP_017921415.1 | 4 - 5 | 1301 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |