Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102171661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695350.3 | XP_005695407.2 | 4 - 5 | 1141 | 65 | |
XM_013973214.2 | XP_013828668.1 | 4 - 5 | 1138 | 65 | ||
GenBank mRNA | JO406198.2 | - | 1 - 2 | 354 | 65 | |
JO406200.2 | - | 1 - 2 | 354 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |