Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044881.1 | XP_017900370.1 | 4 - 5 | 600 | 93 | |
XM_018044882.1 | XP_017900371.1 | 3 - 4 | 734 | 93 | ||
XM_018044883.1 | XP_017900372.1 | 4 - 5 | 600 | 93 | ||
XM_018044884.1 | XP_017900373.1 | 4 - 5 | 600 | 93 | ||
XM_018044885.1 | XP_017900374.1 | 4 - 5 | 600 | 93 | ||
XM_018044886.1 | XP_017900375.1 | 4 - 5 | 600 | 93 | ||
GenBank mRNA | JO428985.2 | - | 3 - 4 | 206 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |