Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3RF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055296.1 | XP_017910785.1 | 3 - 4 | 1872 | 78 | |
XM_018055297.1 | XP_017910786.1 | 3 - 4 | 1872 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |