Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675181.3 | XP_005675238.1 | 5 - 6 | 496 | 60 | |
XM_005675182.3 | XP_005675239.1 | 4 - 5 | 483 | 60 | ||
XM_005675183.3 | XP_005675240.1 | 4 - 5 | 483 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |