Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047497.1 | XP_017902986.1 | 3 - 4 | 1909 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |