Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679817.3 | XP_005679874.1 | 3 - 4 | 321 | 102 | |
XM_005679818.3 | XP_005679875.1 | 3 - 4 | 316 | 102 | ||
XM_018047624.1 | XP_017903113.1 | 3 - 4 | 349 | 102 | ||
GenBank mRNA | JO515757.2 | - | 4 - 5 | 432 | 102 | |
JO598400.2 | - | 4 - 5 | 432 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |