Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013965219.2 | XP_013820673.2 | 2 - 3 | 3791 | 72 | |
GenBank mRNA | JO324065.2 | - | 2 - 3 | 787 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |