Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684173.2 | XP_005684230.2 | 4 - 5 | 786 | 99 | |
GenBank mRNA | KA345176.1 | - | 4 - 5 | 567 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |