Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686525.3 | XP_005686582.1 | 2 - 3 | 452 | 91 | |
XM_018055151.1 | XP_017910640.1 | 1 - 2 | 326 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |