Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688174.3 | XP_005688231.2 | 3 - 4 | 503 | 91 | |
GenBank mRNA | JO631886.2 | - | 1 - 2 | 1496 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |