Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692750.3 | XP_005692807.1 | 4 - 5 | 663 | 84 | |
XM_013971542.2 | XP_013826996.1 | 4 - 5 | 695 | 84 | ||
XM_013971544.2 | XP_013826998.1 | 4 - 5 | 734 | 84 | ||
XM_013971545.2 | XP_013826999.1 | 4 - 5 | 709 | 84 | ||
GenBank mRNA | KA329490.1 | - | 2 - 3 | 972 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |