Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693821.3 | XP_005693878.1 | 2 - 3 | 565 | 71 | |
GenBank mRNA | KA350145.1 | - | 6 - 7 | 1229 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |