Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693821.3 | XP_005693878.1 | 4 - 5 | 878 | 69 | |
GenBank mRNA | KA350145.1 | - | 4 - 5 | 916 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |