Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693821.3 | XP_005693878.1 | 5 - 6 | 1008 | 62 | |
GenBank mRNA | KA350145.1 | - | 3 - 4 | 786 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |