Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693821.3 | XP_005693878.1 | 7 - 8 | 1492 | 86 | |
GenBank mRNA | JO359690.2 | - | 1 - 2 | 318 | 86 | |
KA350145.1 | - | 1 - 2 | 302 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |