Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCWPW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066626.1 | XP_017922115.1 | 9 - 10 | 1363 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |