Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARGFX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005709423.2 | XP_005709480.2 | 1 - 2 | 60 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |