Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTX3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045665.1 | XP_017901154.1 | 3 - 4 | 1954 | 102 | |
GenBank mRNA | JO331665.2 | - | 3 - 4 | 1938 | 102 | |
JO331666.2 | - | 3 - 4 | 1938 | 102 | ||
JO360026.2 | - | 1 - 2 | 622 | 102 | ||
JO396022.2 | - | 2 - 3 | 748 | 102 | ||
JO438312.2 | - | 1 - 2 | 780 | 102 | ||
JO515532.2 | - | 1 - 2 | 408 | 102 | ||
JO521647.2 | - | 1 - 2 | 408 | 102 | ||
JO552976.2 | - | 2 - 3 | 302 | 102 | ||
JO552977.2 | - | 2 - 3 | 302 | 102 | ||
JO562651.2 | - | 1 - 2 | 686 | 102 | ||
JO612967.2 | - | 3 - 4 | 1970 | 102 | ||
KA324292.1 | - | 1 - 2 | 748 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |