Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048475.1 | XP_017903964.1 | 1 - 2 | 273 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |