Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057633.1 | XP_017913122.1 | 5 - 6 | 2231 | 99 | |
XM_018057635.1 | XP_017913124.1 | 6 - 7 | 2466 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |