Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064724.1 | XP_017920213.1 | 2 - 3 | 479 | 86 | |
XM_018064725.1 | XP_017920214.1 | 2 - 3 | 490 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |