Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038204.1 | XP_017893693.1 | 9 - 10 | 2072 | 100 | |
GenBank mRNA | JO427547.1 | - | 3 - 4 | 966 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |