Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039259.1 | XP_017894748.1 | 11 - 12 | 1839 | 66 | |
XM_018039260.1 | XP_017894749.1 | 11 - 12 | 1286 | 66 | ||
XM_018039261.1 | XP_017894750.1 | 10 - 11 | 1387 | 66 | ||
XM_018039262.1 | XP_017894751.1 | 11 - 12 | 1237 | 66 | ||
GenBank mRNA | JO348333.2 | - | 11 - 12 | 1352 | 66 | |
JO410852.2 | - | 11 - 12 | 1246 | 66 | ||
JO410853.2 | - | 11 - 12 | 1246 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |