Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO348517.2 | - | 332 | 78 | ||
JO348518.2 | - | 332 | 78 |