Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680023.3 | XP_005680080.2 | 3 - 4 | 297 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |