Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692161.3 | XP_005692218.3 | 2 - 3 | 504 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |