Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692488.2 | XP_005692545.2 | 8 - 9 | 1129 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |