Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692688.3 | XP_005692745.1 | 2 - 3 | 1056 | 89 | |
GenBank mRNA | KA353246.1 | - | 2 - 3 | 611 | 89 | |
KT288195.1 | - | 2 - 3 | 358 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |