Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FN3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065388.1 | XP_017920877.1 | 4 - 5 | 725 | 88 | |
GenBank mRNA | JO644345.2 | - | 2 - 3 | 574 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |