Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699038.3 | XP_005699095.1 | 7 - 8 | 380 | 82 | |
XM_013975553.2 | XP_013831007.1 | 7 - 8 | 356 | 82 | ||
GenBank mRNA | JO458816.2 | - | 7 - 8 | 344 | 82 | |
JT668021.1 | - | 6 - 7 | 270 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |