Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCF2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047459.1 | XP_017902948.1 | 17 - 18 | 2599 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |