Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM47E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013964737.2 | XP_013820191.2 | 7 - 8 | 1313 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |