Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966567.2 | XP_013822021.2 | 4 - 5 | 585 | 72 | |
GenBank mRNA | JO634801.2 | - | 2 - 3 | 456 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |