Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MATN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055603.1 | XP_017911092.1 | 6 - 7 | 1306 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |