Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055628.1 | XP_017911117.1 | 1 - 2 | 639 | 60 | |
GenBank mRNA | JO646145.2 | - | 1 - 2 | 704 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Capra hircus Probe spans exons |