Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRGQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062526.1 | XP_017918015.1 | 2 - 3 | 662 | 59 | |
XM_018062527.1 | XP_017918016.1 | 2 - 3 | 674 | 59 | ||
GenBank mRNA | JO638626.2 | - | 1 - 2 | 86 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |