Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM86B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063086.1 | XP_017918575.1 | 2 - 3 | 697 | 78 | |
GenBank mRNA | JO615554.2 | - | 1 - 2 | 668 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |