Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065413.1 | XP_017920902.1 | 5 - 6 | 988 | 68 | |
XM_018065414.1 | XP_017920903.1 | 5 - 6 | 989 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |