Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013974207.2 | XP_013829661.2 | 1 - 2 | 2064 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |