Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684096.3 | XP_005684153.1 | 2 - 3 | 280 | 59 | |
GenBank mRNA | JO580733.1 | - | 2 - 3 | 414 | 59 |
Gene Symbol: UNKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | KA341148.1 | - | 619 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |